70104
AQUANAL™-professional Dechlor
determination of zinc; use with : EDTA and copper/zinc
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41113319
Recommended Products
form
solid
measuring range
0.02-1 mg/L
application(s)
environmental
general analytical
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service