Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70068

Supelco

AQUANAL-professional Chlorine HR

determination of chlorine, use with 70070

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41113319

form

tablet

measuring range

5-200 mg/L

application(s)

environmental
general analytical

Packaging

100 tablets

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Oral

target_organs

Thyroid

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service