69820
Millon′s reagent
for the detection of tyrosine
Synonym(s):
Mercury nitrate – nitric acid solution
About This Item
Recommended Products
form
liquid
Quality Level
quality
for the detection of tyrosine
impurities
mercurous nitrate
nitric acid
density
1.4 g/mL at 20 °C
Other Notes
Disclaimer

signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2
supp_hazards
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service