69811
Molecular Weight Marker for proteins
suitable for electrophoresis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
mol wt
Mr 30000-200000
technique(s)
electrophoresis: suitable
suitability
in accordance for electrophoresis test
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Components
SDS-PAGE-Mr:
carbonic anhydrase from bovine erythrocytes: 29000; albumin from chicken egg white: 45000; albumin from bovine serum: 66000; phosphorylase b from rabbit muscle: 97400; β-galactosidase from E. coli: 116000; myosin from rabbit muscle: 205000
carbonic anhydrase from bovine erythrocytes: 29000; albumin from chicken egg white: 45000; albumin from bovine serum: 66000; phosphorylase b from rabbit muscle: 97400; β-galactosidase from E. coli: 116000; myosin from rabbit muscle: 205000
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Eduardo L V Silveira et al.
Clinical and vaccine immunology : CVI, 15(8), 1292-1300 (2008-06-27)
Earlier studies have demonstrated in A/Sn mice highly susceptible to Chagas' disease protective immunity against lethal Trypanosoma cruzi infection elicited by vaccination with an open reading frame (ORF) expressed by amastigotes. In our experiments, we used this mouse model to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service