Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69713

(S)-Oxynitrilase, Manihot esculenta, recombinant from E. coli, CLEA

Synonym(s):

(S)-Oxynitrilase, Manihot esculenta, recombinant from E. coli, cross-linked enzyme aggregate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

recombinant

expressed in E. coli

form

solid

Other Notes

CLEA technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Lopez-Serrano et al.
Biotechnology Letters, 2, 1379-1379 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service