69620
2-Tridecanone
purum, ≥97.0% (GC)
Synonym(s):
Methyl undecyl ketone
About This Item
grade
purum
Quality Level
assay
≥97.0% (GC)
refractive index
n20/D 1.435 (lit.)
bp
134 °C/10 mmHg (lit.)
263 °C (lit.)
mp
24-27 °C (lit.)
density
0.822 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCC(C)=O
InChI
1S/C13H26O/c1-3-4-5-6-7-8-9-10-11-12-13(2)14/h3-12H2,1-2H3
InChI key
CYIFVRUOHKNECG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
224.6 °F - closed cup
flash_point_c
107 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service