Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69575

Sigma-Aldrich

O-Methyl-D-tyrosine

≥99.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
Beilstein/REAXYS Number:
3201704
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.0% (NT)

optical activity

[α]20/D +7.0±1°, c = 1% in 5 M HCl

SMILES string

COc1ccc(C[C@@H](N)C(O)=O)cc1

InChI

1S/C10H13NO3/c1-14-8-4-2-7(3-5-8)6-9(11)10(12)13/h2-5,9H,6,11H2,1H3,(H,12,13)/t9-/m1/s1

InChI key

GEYBMYRBIABFTA-SECBINFHSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ashton Cropp et al.
Nature protocols, 2(10), 2590-2600 (2007-10-20)
The genetic code of living organisms has been expanded to allow the site-specific incorporation of unnatural amino acids into proteins in response to the amber stop codon UAG. Numerous amino acids have been incorporated including photo-crosslinkers, chemical handles, heavy atoms
Jed N Lampe et al.
The Journal of biological chemistry, 285(13), 9594-9603 (2010-01-26)
Cytochrome P450 enzymes are versatile catalysts involved in a wide variety of biological processes from hormonal regulation and antibiotic synthesis to drug metabolism. A hallmark of their versatility is their promiscuous nature, allowing them to recognize a wide variety of
Metabolic engineering. Researchers create first autonomous synthetic life form.
Robert F Service
Science (New York, N.Y.), 299(5607), 640-640 (2003-02-01)
C Lee et al.
Biochemistry and molecular biology international, 31(5), 797-805 (1993-12-01)
An enzyme fraction with catalytic activities for the biosynthesis of the pipecolic acid containing cyclopeptolide SDZ 90-215 was partially purified and characterized from the genus Septoria. The crude cell homogenate was subjected to polyethyleneimine precipitation, ammonium sulfate precipitation and FPLC
S E Shoelson et al.
The Journal of biological chemistry, 264(14), 7831-7836 (1989-05-15)
The receptors for insulin and epidermal growth factor undergo tyrosine autophosphorylation in response to ligand stimulation, while pp60v-src is an unregulated tyrosine kinase. In this report we show that each of the kinases phosphorylates an exogenous peptide that corresponds to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service