Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3ReO3
CAS Number:
Molecular Weight:
249.24
PubChem Substance ID:
UNSPSC Code:
12352300
Beilstein/REAXYS Number:
3927932
MDL number:
grade
purum
assay
≥98.0% (gas-volumetric)
mp
111 °C (lit.), 112-113 °C
SMILES string
C[Re](=O)(=O)=O
InChI
1S/CH3.3O.Re/h1H3;;;;
InChI key
PQTLALPZRPFYIT-UHFFFAOYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
Versatile and efficient catalyst soluble in water and organic solvents; e.g. olefin oxidation, olefin metathesis
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
F.E. Kuhn, W.A. Herrmann
Chemtracts, 14, 59-59 (2001)
W.A. Herrmann et al.
Aqueous-Phase Organomet. Catal., 529-529 (1998)