Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69474

Supelco

3-Ethylmalic acid

≥95.0% (HPLC)

Synonym(s):

β-Ethylmalic acid, (±)-3-Ethyl-2-hydroxybutanedioic acid, 2-Ethyl-3-hydroxysuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
Beilstein/REAXYS Number:
1725256
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥95.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(C(CC)C(O)C(O)=O)=O

InChI

1S/C6H10O5/c1-2-3(5(8)9)4(7)6(10)11/h3-4,7H,2H2,1H3,(H,8,9)(H,10,11)

InChI key

JUCRENBZZQKFGK-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite glyoxalate-condensing enzyme of human pathogens

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Dean et al.
Protein science : a publication of the Protein Society, 5(2), 341-347 (1996-02-01)
The substrate specificity of the NADP-dependent isocitrate dehydrogenase of Escherichia coli was investigated by combining site-directed mutagenesis and utilization of alternative substrates. A comparison of the kinetics of the wild-type enzyme with 2R-malate reveals that the gamma-carboxylate of 2R,3S-isocitrate contributes
K Miyazaki et al.
European journal of biochemistry, 221(3), 899-903 (1994-05-01)
Isocitrate dehydrogenase (ICDH) from an extreme thermophile, Thermus thermophilus HB8, was overexpressed in Escherichia coli. The enzyme was easily purified to homogeneity by a combination of heat treatment (70 degrees C, 20 min) and column chromatography. The N-terminal sequence of
Metabolism of ethylmalic acids by Pseudomonas aeruginosa.
R Rabin et al.
Biochemistry, 7(1), 377-388 (1968-01-01)
A radioactive assay for malate synthase and other glyoxylate condensing enzymes
Wegener, W.S., et al.
Methods in Enzymology, 13, 362-365 (1969)
J R Stern et al.
Journal of bacteriology, 98(1), 147-151 (1969-04-01)
A mutant strain of Salmonella typhimurium (SL 1634 dml-51) capable of growth on d-malate as sole carbon source was shown to produce d-malic enzyme. This enzyme was absent in the parent wild-type strain which was unable to grow on d-malate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service