Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69441

Millipore

Rehydration Fluid for Growth Performance Testing

suitable for microbiology

Synonym(s):

Rehydration Fluid for GPT

Sign Into View Organizational & Contract Pricing

Select a Size

25 X 9 ML
$35.30

$35.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 X 9 ML
$35.30

About This Item

UNSPSC Code:
41106200

$35.30


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

microbiology

storage temp.

2-8°C

Application

Fluid used for the rehydration of microorganisms for growth performance testing.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service