Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69433

Supelco

2-Dehydropantoic acid lithium salt

≥95.0% (HPLC)

Synonym(s):

Ketopantoic acid lithium salt, Lithium 4-hydroxy-3,3-dimethyl-2-oxobutanoate, Lithium 4-hydroxy-3,3-dimethyl-2-oxobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4 · xLi+
Molecular Weight:
146.14 (free acid basis)
Beilstein/REAXYS Number:
2242422
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥95.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(C(C(C)(C)CO)=O)=O

InChI

1S/C6H10O4/c1-6(2,3-7)4(8)5(9)10/h7H,3H2,1-2H3,(H,9,10)

InChI key

PKVVTUWHANFMQC-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite of pantothenate biosynthesis, coenzyme A biosynthesis and in pathways to secondary metabolites.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John W Pinney et al.
Trends in parasitology, 23(11), 548-554 (2007-10-24)
With the completion of sequencing projects for several parasite genomes, efforts are ongoing to make sense of this mass of information in terms of the gene products encoded and their interactions in the growth, development and survival of parasites. The
Yoshiaki Ohashi et al.
Molecular bioSystems, 4(2), 135-147 (2008-01-24)
Metabolic changes in response to histidine starvation were observed in histidine-auxotrophic Escherichia coli using a capillary electrophoresis time-of-flight mass spectrometry (CE-TOFMS)-based metabolomics technique. Prior to the analysis, we prepared an E. coli metabolome list of 727 metabolites reported in the
D R Wilken et al.
The Journal of biological chemistry, 250(6), 2311-2314 (1975-03-25)
The stereospecificity of hydrogen transfer from NADPH to the appropriate carbonyl substrate catalyzed by ketopantoic acid and ketopantoyl acid and ketopantoyl lactone reductases of yeast (Saccharomyces cerevisiae) and Escherichia coli has been determined. Yeast and E. coli ketopantoic acid reductases
Hiroya Tomita et al.
Molecular microbiology, 90(2), 307-321 (2013-08-15)
Coenzyme A (CoA) biosynthesis in bacteria and eukaryotes is regulated primarily by feedback inhibition towards pantothenate kinase (PanK). As most archaea utilize a modified route for CoA biosynthesis and do not harbour PanK, the mechanisms governing regulation of CoA biosynthesis
Cheryl N Miller et al.
Journal of bacteriology, 195(5), 965-976 (2012-12-18)
Pantothenate, commonly referred to as vitamin B(5), is an essential molecule in the metabolism of living organisms and forms the core of coenzyme A. Unlike humans, some bacteria and plants are capable of de novo biosynthesis of pantothenate, making this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service