Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69407

Supelco

GMO Genomic DNA Standard Set for Maize NK603, GA21 and CBH-351 "Starlink"

analytical standard

Synonym(s):

Maize GMO Standard Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

suitability

in accordance for tests for cross contaminations

application(s)

cleaning products
cosmetics
food and beverages
microbiology
personal care

format

neat

storage temp.

2-8°C

Application

GMO Genomic DNA Standard Set for Maize NK603, GA21 and CBH-351 "Starlink" is used as a genetically modified reference material for the detection of the analyte:
  • In a multiplex real-time PCR reaction using the food-proof GMO screening kit.[1]
  • In food products using pentaplex realtime PCR systems.[2]

Analysis Note

real-time PCR test: corresponds;
each tube contains maize DNA containing approximately 1% genet. modified DNA

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Five pentaplex real-time PCR systems for the efficient determination of 20 genetically modified maize traits in food
Koppel R, et al.
European Food Research and Technology, 243(2), 215-225 (2017)
Development of a qualitative, multiplex real-time PCR kit for screening of genetically modified organisms (GMOs)
Dorries H-H, et al.
Analytical and Bioanalytical Chemistry, 396(6), 2043-2054 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service