69262
mono-Cyclohexyl phthalate
analytical standard
Synonym(s):
1,2-Benzenedicarboxylic acid 1-cyclohexyl ester, Phthalic acid monocyclohexyl ester
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
95-99 °C
application(s)
environmental
format
neat
storage temp.
2-8°C
SMILES string
O=C(OC1CCCCC1)C2=C(C(O)=O)C=CC=C2
InChI
1S/C14H16O4/c15-13(16)11-8-4-5-9-12(11)14(17)18-10-6-2-1-3-7-10/h4-5,8-10H,1-3,6-7H2,(H,15,16)
InChI key
PMDKYLLIOLFQPO-UHFFFAOYSA-N
Application
Recommended products
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service