Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69207

1-Heptanesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6SO2Cl
CAS Number:
Molecular Weight:
198.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CCCCCCCS(Cl)(=O)=O

InChI

1S/C7H15ClO2S/c1-2-3-4-5-6-7-11(8,9)10/h2-7H2,1H3

InChI key

HXIUMEKDVYLMOK-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service