Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69091

Supelco

Dinotefuran

reference material

Synonym(s):

(RS)-N-Methyl-N′-nitro-N″-[(tetrahydro-3-furanyl)methyl]guanidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N4O3
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

CN\C(N[N+]([O-])=O)=N/CC1CCOC1

InChI

1S/C7H14N4O3/c1-8-7(10-11(12)13)9-4-6-2-3-14-5-6/h6H,2-5H2,1H3,(H2,8,9,10)

InChI key

YKBZOVFACRVRJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dinotefuran, a new neonicotinoid insecticide has low mammalian toxicity and great insecticidal activity against a broad range of pests

Find more information here: Neonicotinoids
This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service