Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69038

Supelco

Conductivity Standard

conductivity 350000 μS/cm at 25 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

20-50% KCl (T)

technique(s)

affinity chromatography: suitable

conductivity

346500-353500 μS/cm at 25 °C

density

1.133-2.421 g/cm3 at 20 °C

suitability

suitable for chromatography

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

standard

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2 - STOT RE 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service