Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68709

Supelco

3′,4′-Dimethoxyacetophenone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCH3
CAS Number:
Molecular Weight:
180.20
Beilstein/REAXYS Number:
781213
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

bp

286-288 °C (lit.)

mp

47-54 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccc(cc1OC)C(C)=O

InChI

1S/C10H12O3/c1-7(11)8-4-5-9(12-2)10(6-8)13-3/h4-6H,1-3H3

InChI key

IQZLUWLMQNGTIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service