Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68558

Supelco

2-Ethylpyridine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein/REAXYS Number:
106480
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.495-1.501
n20/D 1.496 (lit.)

bp

149 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCc1ccccn1

InChI

1S/C7H9N/c1-2-7-5-3-4-6-8-7/h3-6H,2H2,1H3

InChI key

NRGGMCIBEHEAIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylpyridine, a nitrogen aromatic compound, is grouped under the class of azaarenes. It is generally found as a component of fuels and combustion products.[1][2]

Application

2-Ethylpyridine may be used as an analytical standard for the determination of the analyte in mainstream cigarette smoke,[3] solvent refined coal (SRC) recycle oil,[1] and aromatic base mixture from coal gasification process[2] by various chromatography based techniques.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capillary gas chromatography/mass spectrometric determination of nitrogen aromatic compounds in complex mixtures
Novotny M, et al.
Analytical Chemistry, 52(3), 401-406 (1980)
Isolation of mutagenic aromatic amines from a coal conversion oil by cation exchange chromatography
Haugen DA, et al.
Analytical Chemistry, 54(1), 32-37 (1982)
N P Kulshreshtha et al.
Journal of chromatography. A, 985(1-2), 303-312 (2003-02-13)
A new technique has been developed for the quantitative analysis of pyridines in mainstream cigarette smoke using a GC-MS technique. For analysis, 10 cigarettes are smoked using conditions based on US Federal Trade Commission recommendations. The smoke is collected in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service