Skip to Content
MilliporeSigma

68512

4-Methyl-1-pentene

purum, ≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2CHCH2CH=CH2
CAS Number:
Molecular Weight:
84.16
EC Number:
211-720-1
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1731096
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

>1 (vs air)

vapor pressure

4.45 psi ( 20 °C)

grade

purum

assay

≥97.0% (GC)

autoignition temp.

572 °F

impurities

isomeric hexens

refractive index

n20/D 1.382 (lit.), n20/D 1.383

bp

53-54 °C (lit.)

density

0.665 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC=C

InChI

1S/C6H12/c1-4-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

WSSSPWUEQFSQQG-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 4-Methyl-1-pentene




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library