Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68510

Supelco

4-Methyl-1-pentene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein/REAXYS Number:
1731096
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

4.45 psi ( 20 °C)

assay

≥99.5% (GC)

autoignition temp.

572 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.382 (lit.)
n20/D 1.382

bp

53-54 °C (lit.)

density

0.665 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

SMILES string

CC(C)CC=C

InChI

1S/C6H12/c1-4-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

WSSSPWUEQFSQQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-23.8 °F

flash_point_c

-31 °C

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service