Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68458

Supelco

Nitrofen-(nitrophenyl-d4)

PESTANAL®, analytical standard

Synonym(s):

2,4-Dichlorophenyl 4-nitrophenyl-d3 ether, Nitrofen-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D4H3Cl2NO3
Molecular Weight:
288.12
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥95 atom % D

product line

PESTANAL®

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

ClC(C=C(Cl)C=C1)=C1OC2=C([2H])C([2H])=C([N+]([O-])=O)C([2H])=C2[2H]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service