Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3C6H3(NO2)NH2
CAS Number:
Molecular Weight:
152.15
EC Number:
209-329-6
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
1868029
grade
purum
assay
≥99.0% (GC)
mp
93-96 °C
SMILES string
Cc1cccc(c1N)[N+]([O-])=O
InChI
1S/C7H8N2O2/c1-5-3-2-4-6(7(5)8)9(10)11/h2-4H,8H2,1H3
InChI key
FCMRHMPITHLLLA-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service