Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67891

Sigma-Aldrich

3-Deoxy-D-manno-octulosonate Aldolase from E. coli

≥1.0 U/mg

Synonym(s):

2-Keto-3-deoxyoctonate Aldolase, KDO Aldolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Escherichia coli

Quality Level

form

powder

specific activity

≥1.0 U/mg

storage temp.

−20°C

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 nmol 2-Keto-3-deoxyoctonate per hour at pH 7.0 and 37°C.

Other Notes

Catalyst for stereoselective aldol reactions[1][2][3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Sugai et al.
Journal of the American Chemical Society, 115, 413-413 (1993)
The biosynthesis of cell wall lipopolysaccharide in Escherichia coli. V. Purification and properties of 3-deoxy-D-manno-octulosonate aldolase.
M A Ghalambor et al.
The Journal of biological chemistry, 241(13), 3222-3227 (1966-07-10)
B.R. Knappmann et al.
Applied Microbiology and Biotechnology, 33, 324-324 (1990)
U. Kragl et al.
Angewandte Chemie (International Edition in English), 103, 854-854 (1991)

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service