Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67790

Supelco

Cinoxate

analytical standard

Synonym(s):

2-Ethoxyethyl 3-(4-methoxyphenyl)-2-propenoate, 2-Ethoxyethyl 4-methoxycinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O4
CAS Number:
Molecular Weight:
250.29
Beilstein/REAXYS Number:
2728232
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCOCCOC(=O)\C=C\c1ccc(OC)cc1

InChI

1S/C14H18O4/c1-3-17-10-11-18-14(15)9-6-12-4-7-13(16-2)8-5-12/h4-9H,3,10-11H2,1-2H3/b9-6+

InChI key

CMDKPGRTAQVGFQ-RMKNXTFCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cinoxate belongs to the group of cinnamates.It is widely used as a sunscreen agent in cosmetic products due to its ability to absorb ultraviolet B (UVB) rays.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sunscreen agents in cosmetic products by gas chromatography and gas chromatography?mass spectrometry
Ikeda D, et al.
Journal of Chromatography A, 513, 321- 326 (1990)
Rigel.SD, et al.
Photoaging (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service