Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67609

Supelco

1,2-Dibromo-3-chloropropane

reference material

Synonym(s):

DBCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCHBrCH2Cl
CAS Number:
Molecular Weight:
236.33
Beilstein/REAXYS Number:
1732077
MDL number:
UNSPSC Code:
41116107
Product 67609 is not currently sold in your country. Contact Technical Service

grade

reference material

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n/D 1.5542

bp

60-65 °C/7.5 mmHg

density

2.081 g/mL at 25 °C

format

neat

storage temp.

2-8°C

SMILES string

ClCC(Br)CBr

InChI

1S/C3H5Br2Cl/c4-1-3(5)2-6/h3H,1-2H2

InChI key

WBEJYOJJBDISQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 1B - Repr. 1A - STOT RE 2 Oral

target_organs

Testes

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

170.1 °F

flash_point_c

76.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles A Easley et al.
Stem cell research, 14(3), 347-355 (2015-04-13)
Environmental influences and insults by reproductive toxicant exposure can lead to impaired spermatogenesis or infertility. Understanding how toxicants disrupt spermatogenesis is critical for determining how environmental factors contribute to impaired fertility. While current animal models are available, understanding of the
Dane R Letourneau et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 14(11), 2108-2118 (2015-10-07)
The photochemically mediated dechlorination of polyhalogenated compounds represents a potential decontamination strategy and a relevant environmental process in chemically reducing media. We report the UV irradiation of natural and artificial waters containing natural dissolved organic matter to effect the photo-sensitized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service