67402
Light Green SF Yellowish
analytical standard
Synonym(s):
Acid Green 5
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥97.0% (TLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤4.0% water
mp
288 °C (dec.) (lit.)
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)\C(=C3\C=CC(\C=C3)=[N+](/CC)Cc4cccc(c4)S([O-])(=O)=O)c5ccc(cc5)S([O-])(=O)=O
InChI
1S/C37H36N2O9S3.2Na/c1-3-38(25-27-7-5-9-35(23-27)50(43,44)45)32-17-11-29(12-18-32)37(31-15-21-34(22-16-31)49(40,41)42)30-13-19-33(20-14-30)39(4-2)26-28-8-6-10-36(24-28)51(46,47)48;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);;/q;2*+1/p-2
InChI key
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Caution
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service