Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

67389

Supelco

Hexyl chloroformate

derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2(CH2)5CH3
CAS Number:
Molecular Weight:
164.63
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (GC derivatization)

Quality Level

vapor pressure

0.06 psi ( 20 °C)

assay

≥97.0% (GC)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)

bp

60-61 °C/7 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCOC(Cl)=O

InChI

1S/C7H13ClO2/c1-2-3-4-5-6-10-7(8)9/h2-6H2,1H3

InChI key

KIWBRXCOTCXSSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl chloroformate belongs to the class of chloroformates.[1] It is widely used in the derivatization of highly hydrophilic compounds to hexyl esters, carbonates and carbamates.[2]

Application

Hexyl chloroformate may be used as a derivatizing reagent for the quantification of antiepileptic drugs,[3] cathinones, amphetamine-like stimulants and other new psychoactive substances[4] in biological samples using gas chromatography coupled to mass spectrometry (GC-MS).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabapentin, Pregabalin and Vigabatrin quantification in human serum by GC--MS after hexyl chloroformate derivatization
Hlozek T, et al.
Journal of Analytical Toxicology, 40(9), 749-753 (2016)
Stacy D Brown et al.
Forensic science international, 171(2-3), 142-150 (2006-12-13)
A solid-phase microextraction-gas chromatographic-mass spectrometric (SPME-GC-MS) method has been developed and validated for measuring four club drugs in human urine. These drugs include gamma-hydroxybutyrate (GHB), ketamine (KET), methamphetamine (MAMP), and methylenedioxymethamphetamine (MDMA). These drugs are referred to as 'club drugs'
Chloroformates in gas chromatography as general purpose derivatizing agents
Husek P
Journal of Chromatography. B, Biomedical Sciences and Applications, 717(1-2), 57-91 (1998)
Rapid and simple procedure for the determination of cathinones, amphetamine-like stimulants and other new psychoactive substances in blood and urine by GC--MS
Mercieca G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 149, 494-501 (2018)
Determination of trace amounts of highly hydrophilic compounds in water by direct derivatization and gas chromatography?mass spectrometry
Minero C, et al.
Fresenius Journal of Analytical Chemistry, 350(6), 403-409 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service