Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67359

Supelco

Tectorigenin

analytical standard

Synonym(s):

4′,5,7-Trihydroxy 6-methoxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxy-phenyl)-6-methoxy-chromen-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 5-Hydroxyglycitein, Tectrigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Beilstein/REAXYS Number:
305601
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

suitability

conforms to structure for Infrared spectrum

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC(C=C1)=CC=C1C2=COC3=CC(O)=C(OC)C(O)=C3C2=O

InChI

1S/C16H12O6/c1-21-16-11(18)6-12-13(15(16)20)14(19)10(7-22-12)8-2-4-9(17)5-3-8/h2-7,17-18,20H,1H3

InChI key

OBBCRPUNCUPUOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service