Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67344

Supelco

Indoxacarb

reference material

Synonym(s):

Methyl 7-chloro-2,5-dihydro-2-[N-(methoxycarbonyl)-4-(trifluoromethoxy)anilinocarbonyl]indeno[1,2-e][1,3,4]oxadiazine-4a(3H)carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClF3N3O7
CAS Number:
Molecular Weight:
527.83
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

COC(=O)N(C(=O)N1CO[C@]2(Cc3cc(Cl)ccc3C2=N1)C(=O)OC)c4ccc(OC(F)(F)F)cc4

InChI

1S/C22H17ClF3N3O7/c1-33-18(30)21-10-12-9-13(23)3-8-16(12)17(21)27-28(11-35-21)19(31)29(20(32)34-2)14-4-6-15(7-5-14)36-22(24,25)26/h3-9H,10-11H2,1-2H3/t21-/m0/s1

InChI key

VBCVPMMZEGZULK-NRFANRHFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 1

target_organs

Heart,Nervous system,Blood system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service