This enrichment broth enables the reactivation of damaged Salmonella from dehydrated media. The supplementation of ferrioxamine E significantly improves the recovery of Salmonella from contaminated foods.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Viable but nonculturable (VBNC) bacteria describes cells that cannot be normally cultured however, “resuscitation” of these cells can be mediated by a physical and different kind of chemical stimuli.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.