Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67318

Supelco

N-(2-Maleimidoethyl)ferrocenecarboxamide

derivatization grade (HPLC), ≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16FeN2O3
CAS Number:
Molecular Weight:
352.17
MDL number:
UNSPSC Code:
23151816
PubChem Substance ID:

grade

derivatization grade (HPLC)

assay

≥98.0% (HPLC)

form

solid

composition

carbon, 56.8-59.1% CHN

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.O=C(NCCN2C(=O)C=CC2=O)[C]3[CH][CH][CH][CH]3

InChI

1S/C12H11N2O3.C5H5.Fe/c15-10-5-6-11(16)14(10)8-7-13-12(17)9-3-1-2-4-9;1-2-4-5-3-1;/h1-6H,7-8H2,(H,13,17);1-5H;

InChI key

QTEUVLCRCJSEAQ-UHFFFAOYSA-N

Application

Derivatisation reagent for thiols and disulfides by LC/MS/MS

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bettina Seiwert et al.
Analytical chemistry, 79(18), 7131-7138 (2007-08-21)
A method for the simultaneous determination of a series of thiols and disulfides in urine samples has been developed based on the sequential labeling of free and bound thiol functionalities with two ferrocene-based maleimide reagents. The sample is first exposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service