Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66373

Supelco

Micro particles based on silicon dioxide

size: 3 μm

Synonym(s):

Silicon dioxide microsphere (3 μm), Micro silicon (3 μm), Beads based on silicon dioxide, microsize, Silicon dioxide beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

grade

analytical standard

Quality Level

form

aqueous suspension

concentration

5% (solids)

particle size

3 μm std dev ≤0.25 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ideal for profiling particle size distribution (PSD) of a particle system.
The particle size of 3 μm is characterized using Coulter multisizer III.

Application

Silicon dioxide beads, 3 μm have a wide range of uses from use as a flow agent to electrical and medical applications.
Used for regular calibration and monitoring of particle size analyzers.
Also used in microfluidic devices to:
  • track flows induced by enzyme micropumps[1]
  • investigate the effects of pumps′ activity on silica tracer microparticles near solid-liquid interface.[2]

Features and Benefits

  • suitable for routine instrument calibration checking, testing and corrections
  • particle size traceable to Community Bureau of Reference (BCR) standards
  • available in 5 mL pack size as a neat sample

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Glucose Oxidase Micropumps: Multi-Faceted Effects of Chemical Activity on Tracer Particles near the Solid-Liquid Interface
Munteanu RE, et al.
Chemosphere, 4(3), 73-73 (2019)
The impact of geometrical confinement in a slab on the behavior of tracer particles near active glucose oxidase micropump
Munteanu RE, et al.
Coll. Polymer Sci., 299(2), 297-306 (2021)
Surachate Kalasin et al.
Langmuir : the ACS journal of surfaces and colloids, 26(4), 2317-2324 (2010-02-10)
This paper explores the particle-level dynamics involved in the capture of gently flowing microparticles on adhesive planar surfaces, governed by electrostatic interactions. The work focuses on conditions which produce sustained microparticle rolling, useful for the development of microfluidic devices which
Jason S Kim et al.
Analytical and bioanalytical chemistry, 398(6), 2373-2382 (2010-06-08)
Micron-sized particles have primarily been used in microfabricated flow cytometers for calibration purposes and proof-of-concept experiments. With increasing frequency, microparticles are serving as a platform for assays measured in these small analytical devices. Light scattering has been used to measure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service