Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66324

Supelco

Eupalitin

analytical standard

Synonym(s):

3,4′,5-Trihydroxy-6,7-dimethoxyflavone, 3,5-Dihydroxy-2-(4-hydroxyphenyl)-6,7-dimethoxy-4H-1-benzopyran-4-one, 3,5-Dihydroxy-2-(4-hydroxyphenyl)-6,7-dimethoxychromen-4-one, Betuletol, Eupaletin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein/REAXYS Number:
1354525
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O=C1C(O)=C(C2=CC=C(O)C=C2)OC3=CC(OC)=C(OC)C(O)=C31

InChI

1S/C17H14O7/c1-22-11-7-10-12(14(20)17(11)23-2)13(19)15(21)16(24-10)8-3-5-9(18)6-4-8/h3-7,18,20-21H,1-2H3

InChI key

KWMAWXWUGIEVDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarjeel Kaleem et al.
Cell biology international, 40(2), 196-203 (2015-10-24)
Prostate cancer is the second most common malignancy in the human reproductive system. Eupalitin is one of the O-methylated flavonol-exhibited enhanced cancer chemopreventive agents. The current study highlights the structural determination of eupalitin and aims to explore the antitumor activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service