Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65819

Supelco

Acrolein O-pentafluorophenylmethyl-oxime

analytical standard

Synonym(s):

Acrolein O-2,3,4,5,6-PFBHA-oxime, Propenal O-pentafluorophenylmethyl-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F5NO
CAS Number:
Molecular Weight:
251.15
Beilstein/REAXYS Number:
9270059
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(CO\N=C\C=C)c(F)c1F

InChI

1S/C10H6F5NO/c1-2-3-16-17-4-5-6(11)8(13)10(15)9(14)7(5)12/h2-3H,1,4H2/b16-3+

InChI key

ICDUEGOPUWNJNF-HQYXKAPLSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service