Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65480

Sigma-Aldrich

4′-Methylacetophenone

purum, ≥96.0% (GC)

Synonym(s):

Methyl p-tolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
Beilstein/REAXYS Number:
606053
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥96.0% (GC)

refractive index

n20/D 1.533 (lit.)
n20/D 1.535

bp

226 °C (lit.)

mp

22-24 °C (lit.)

density

1.004 g/mL at 20 °C
1.005 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccc(C)cc1

InChI

1S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6H,1-2H3

InChI key

GNKZMNRKLCTJAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Uwai et al.
Bioorganic & medicinal chemistry, 16(3), 1084-1089 (2007-11-17)
Stereoselective reductive metabolism of various p-substituted acetophenone derivatives was studied using isolated rat liver 3alpha-hydroxysteroid dehydrogenase (3alpha-HSD). Kinetic experiments were performed and analyzed by measuring the products by HPLC using a chiral column. The results demonstrated that the presence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service