Skip to Content
MilliporeSigma

64770

2-Methoxybenzaldehyde

purum, ≥98.0% (GC)

Synonym(s):

o-Anisaldehyde, Salicylaldehyde methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
EC Number:
205-171-7
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
606301
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.0% (GC)

impurities

≤2% 2-methoxybenzoic acid

bp

238 °C (lit.)

mp

34-40 °C (lit.), 35-37 °C

density

1.127 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccccc1OC

InChI

1S/C8H8O2/c1-10-8-5-3-2-4-7(8)6-9/h2-6H,1H3

InChI key

PKZJLOCLABXVMC-UHFFFAOYSA-N

Biochem/physiol Actions

Naturally-occurring aromatic aldehyde with acaricidal activity. May condense with L-tryptophan in foods to form the corresponding phenolic tetrahydro-β-carboline-3-carboxylic acid which is a potent antioxidant.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly



Global Trade Item Number

SKUGTIN
CDS001551-250MG04061828965298