Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64710

Sigma-Aldrich

4′-Methoxyacetophenone

purum, ≥99.0% (GC)

Synonym(s):

4-Acetylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCH3
CAS Number:
Molecular Weight:
150.17
Beilstein/REAXYS Number:
742313
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥99.0% (GC)

form

solid

bp

152-154 °C/26 mmHg (lit.)

mp

36-38 °C (lit.)
37-39 °C

SMILES string

COc1ccc(cc1)C(C)=O

InChI

1S/C9H10O2/c1-7(10)8-3-5-9(11-2)6-4-8/h3-6H,1-2H3

InChI key

NTPLXRHDUXRPNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Toxicological characteristics of acetylanisole].
M I Makaruk et al.
Gigiena i sanitariia, (4)(4), 86-87 (1985-04-01)
Wen-Yong Lou et al.
Journal of biotechnology, 143(3), 190-197 (2009-07-21)
The biocatalytic enantioselective reduction of 4'-methoxyacetophenone to (S)-1-(4-methoxyphenyl)ethanol was successfully conducted in a hydrophilic IL-containing co-solvent system using immobilized Rhodotorula sp. AS2.2241 cells. Of all the tested ILs, the best results were observed with the novel IL 1-(2'-hydroxy)ethyl-3-methylimidazolium nitrate (C(2)OHMIM.NO(3))
Sanja Ćavar et al.
Pharmaceutical biology, 48(2), 170-176 (2010-07-22)
Stachys menthifolia Vis. (Lamiaceae) is an endemic species from the Balkan Peninsula spread throughout Albania, Greece, Montenegro, and Croatia. This article presents the first investigation of the essential oil composition of this species from Croatia. Aerial parts of the plant
Shakila K Evans et al.
Chemical biology & drug design, 75(3), 333-337 (2010-07-28)
Acetophenones were screened for activity against positive phototaxis of Chlamydomonas cells, a process that requires co-ordinated flagellar motility. The structure-activity relationships of a series of acetophenones are reported, including acetophenones that affect flagellar motility and cell viability. Notably, 4-methoxyacetophenone, 3,4-dimethoxyacetophenone
Shinya Tachibana et al.
Journal of oleo science, 57(2), 107-113 (2008-01-17)
The synthesis and physiological activity of thiophenes and furans with methoxyacetophenone derivatives were examined. 3-Methoxyacetophenone (1) and 4-methoxyacetophenone (2) were converted, respectively, to the oximes (3) and (4) by oximation with hydroxylamine hydrochloride, and to the primary amines (5) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service