Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64260

Sigma-Aldrich

Methanesulfonyl chloride

puriss., ≥99.0% (AT)

Synonym(s):

Mesyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2Cl
CAS Number:
Molecular Weight:
114.55
Beilstein/REAXYS Number:
506297
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (AT)

refractive index

n20/D 1.452 (lit.)
n20/D 1.452

bp

60 °C/21 mmHg (lit.)

density

1.48 g/mL at 25 °C (lit.)

SMILES string

CS(Cl)(=O)=O

InChI

1S/CH3ClO2S/c1-5(2,3)4/h1H3

InChI key

QARBMVPHQWIHKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Connor et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 3(11), 2164-2171 (1983-11-01)
Excitatory postsynaptic currents (EPSCs), miniature excitatory postsynaptic currents (MEPSCs), and acetylcholine-induced current fluctuations (noise) have been studied in voltage-clamped bullfrog parasympathetic ganglion cells of the atrial septum. EPSCs were also recorded from voltage-clamped sympathetic B cells and, in general, it
R Kullberg et al.
The Journal of physiology, 374, 413-427 (1986-05-01)
The development of miniature end-plate currents (m.e.p.c.s) was studied in the superior oblique and interhyoideus muscles of Xenopus laevis. An analysis of m.e.p.c. decays shows that each muscle possesses its own characteristic programme of end-plate current development. In the superior
Zdzisław Brzozowski et al.
Journal of medicinal chemistry, 45(2), 430-437 (2002-01-11)
A new series of 8-chloro-5,5-dioxoimidazo[1,2-b][1,4,2]benzodithiazine derivatives 8-25 with heteroaryloxycarbonyl or heteroarylcarbamoyl substituents at position 7 have been synthesized as potential antitumor agents. In this procedure a novel type of mixed anhydride 7 was prepared from 8-chloro-5,5-dioxoimidazo[1,2-b][1,4,2]benzodithiazine-7-carboxylic acid 6 and methanesulfonyl
Hiroshi Nagase et al.
The Journal of organic chemistry, 73(20), 8093-8096 (2008-09-25)
The treatment of morphinan 1 with NaH and MsCl provided very stable iminium salt 7 possessing propellane skeleton. One of the synthesized iminium salts 7, isobutyl derivative 7b, was crystallized and its structure was determined by X-ray crystallography. The natural
Maurizio Tamba et al.
Journal of the American Chemical Society, 129(28), 8716-8723 (2007-06-22)
The one-electron reduction of methanesulfonyl chloride (MeSO2Cl) leads, in the first instance, to an electron adduct MeSO2Cl(.)(-) which lives long enough for direct detection and decays into sulfonyl radicals MeSO2(.) and Cl(-), with k = 1.5 x 10(6) s(-1). Both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service