Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64220

Sigma-Aldrich

2-Methyl-2-propen-1-ol

purum, ≥98.0% (GC)

Synonym(s):

β-Methallyl alcohol, ‘Isobutenol’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)CH2OH
CAS Number:
Molecular Weight:
72.11
Beilstein/REAXYS Number:
969226
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

refractive index

n20/D 1.426 (lit.)
n20/D 1.426

bp

113-115 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

CC(=C)CO

InChI

1S/C4H8O/c1-4(2)3-5/h5H,1,3H2,2H3

InChI key

BYDRTKVGBRTTIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

89.6 °F - closed cup

flash_point_c

32 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Bittman et al.
Chemistry and physics of lipids, 50(2), 99-103 (1989-05-01)
The synthesis of C2-methyl-1,2-di-O-hexadecylglycerophosphocholine from methylallyl alcohol is described and the 13C-NMR assignments of all glycerol backbone and headgroup resonances are provided. Bond rotations in the glycerol backbone of this diether phospholipid derivative are impeded by steric hindrance exerted by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service