Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64208

Supelco

3-(Trimethoxysilyl)propyl methacrylate

Selectophore

Synonym(s):

Silane A174, [3-(Methacryloyloxy)propyl]trimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
248.35
Beilstein/REAXYS Number:
1952435
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

assay

≥95.0%

form

liquid

refractive index

n20/D 1.431 (lit.)

bp

190 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCOC(=O)C(C)=C)(OC)OC

InChI

1S/C10H20O5Si/c1-9(2)10(11)15-7-6-8-16(12-3,13-4)14-5/h1,6-8H2,2-5H3

InChI key

XDLMVUHYZWKMMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Atai et al.
Dental materials : official publication of the Academy of Dental Materials, 28(2), 133-145 (2011-12-06)
The study evaluates properties of an experimental dental composite consisting of a porous thermally sintered nano-silica as filler. The properties are compared with those of an experimental composite containing micro fillers and a commercially available nano-composite, Filtek Supreme(®) Translucent. Different
Caroline Cvetkovic et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(28), 10125-10130 (2014-07-02)
Combining biological components, such as cells and tissues, with soft robotics can enable the fabrication of biological machines with the ability to sense, process signals, and produce force. An intuitive demonstration of a biological machine is one that can produce
Jiangdong Dai et al.
Journal of hazardous materials, 205-206, 179-188 (2012-01-21)
In the work, we reported an effective method for the preparation of molecularly imprinted nanoparticles with superparamagnetic susceptibility through atom transfer radical emulsion polymerization (ATREP), and then as-prepared magnetic molecularly imprinted nanoparticles (MMINs) were evaluated as adsorbents for selective recognition
Danqing Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(13), 3880-3885 (2015-03-17)
In this work we propose randomly ordered polydomain nematic liquid crystal polymer networks to reversibly generate notable jagged relief patterns at a polymer coating surface by light illumination. The domain size is controlled by the addition of traces of partly
Christie Ying Kei Lung et al.
Dental materials : official publication of the Academy of Dental Materials, 28(8), 863-872 (2012-06-05)
To evaluate and compare three novel coupling agents: 2-hydroxyethyl methacrylate, itaconic acid and oleic acid to two silane coupling agents, one commercial silane product and 3-acryloxypropyltrimethoxysilane on the bond durability of resin composite to zirconia. Zirconia samples were silica-coated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service