Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63330

Sigma-Aldrich

Diethyl malonate

purum, ≥97.0% (GC)

Synonym(s):

Malonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
774687
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.52 (vs air)

vapor pressure

1 mmHg ( 40 °C)

grade

purum

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.413 (lit.)

bp

199 °C (lit.)

mp

−51-−50 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3

InChI key

IYXGSMUGOJNHAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renhua Fan et al.
The Journal of organic chemistry, 72(15), 5905-5907 (2007-06-26)
A highly efficient, one-flask tandem Knoevenagel-Michael addition reaction of sulfonimines with diethyl malonate in the presence of a catalytic amount of base affords the corresponding arylidene dimalonates in good to excellent yields.
Chunying Shu et al.
Journal of the American Chemical Society, 129(50), 15710-15717 (2007-11-22)
The first reactions of trimetallic nitride templated endohedral metallofullerenes (TNT EMFs) with carbon radicals generated from diethyl malonate catalyzed by manganese(III) acetate are reported. Two methano monoadducts, Sc3N@C80-A and Sc3N@C80-B, were isolated and characterized. Sc3N@C80-A contains two ester moieties, whereas
Shigeyuki Yamada et al.
Organic & biomolecular chemistry, 5(9), 1442-1449 (2007-04-28)
The addition of heteroaromatic lithium reagents 2 to a THF solution of perfluorocyclopentene (1) provided preferentially the corresponding monosubstituted products 5, while the addition of 1 to 2 effectively gave the 1,2-disubstituted products 6 in good to excellent yields. The
Sau Fan Yip et al.
Organic letters, 9(17), 3469-3472 (2007-07-20)
An effective method in targeting alpha-aryl malonates is reported. In the presence of a catalytic amount of 2-picolinic acid and CuI, the coupling of aryl iodides with diethyl malonate proceeds smoothly even at room temperature. The high levels of functional
Hua Li et al.
Nucleosides, nucleotides & nucleic acids, 29(8), 581-590 (2010-07-28)
This article describes a very simple route for synthesizing novel lipophilic phosphonate bis(t-bu-SATE) prodrugs of acyclic cyclopentenylated nucleosides such as adenine 17 and cytosine 18. The key intermediate 6 was constructed via a ring-closing metathesis of compound 5, which could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service