Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631825

Supelco

Ammonium-15N nitrate-15N solution

suitable for NMR (reference standard), 98 atom % 15N, 66.67 mg/mL in D2O (99 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15NH415NO3
Molecular Weight:
82.03
MDL number:
UNSPSC Code:
12142201
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

98 atom % 15N

concentration

66.67 mg/mL in D2O (99 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

mass shift

M+2

Features and Benefits

15N sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

pictograms

Flame over circle

signalword

Warning

hcodes

Hazard Classifications

Ox. Liq. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service