Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63142

Sigma-Aldrich

Magnesium sulfate heptahydrate

purum p.a., ≥99.0% (T)

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.0% (T)

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

grade

purum p.a.

assay

≥99.0% (T)

pH

5.0-8.0 (25 °C, 50 mg/mL in H2O)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ESR studies of (Cu(H2O)6)2+ in magnesium sulfate heptahydrate.
Radhakrishna S and Bhaskar Rao T.
Journal of Magnetic Resonance, 32(1), 71-81 (1978)
Sulfates on Mars: A systematic Raman spectroscopic study of hydration states of magnesium sulfates.
Wang A, et al.
Geochimica et Cosmochimica Acta, 70(24), 6118-6135 (2006)
Ran D Goldman et al.
Pediatrics, 132(6), e1634-e1641 (2013-11-28)
Vaso-occlusive episodes (VOEs) are the most common complication of sickle cell disease in children. Treatment with magnesium seems to improve cellular hydration and may result in reduced vaso-occlusion. This study aimed to determine if intravenous (IV) magnesium sulfate (MgSO4) reduces
Oluwakemi Badaki-Makun et al.
Pediatric blood & cancer, 61(6), 1049-1054 (2014-01-21)
Multiple recent Sickle Cell Disease studies have been terminated due to poor enrollment. We developed methods to overcome past barriers and utilized these to study the efficacy and safety of intravenous magnesium for vaso-occlusive crisis (VOC). We describe the methods
E S Bain et al.
BJOG : an international journal of obstetrics and gynaecology, 121(5), 595-603 (2014-01-07)
To evaluate a slower (compared with a standard) infusion rate of the loading dose of magnesium sulphate for preterm fetal neuroprotection as a strategy to reduce maternal adverse effects. Randomised controlled trial. South Australian maternity hospital. Fifty-one women at <30

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service