Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62355

Millipore

Listeria Selective Agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

solid

shelf life

limited shelf life, expiry date on the label

composition

agar, 13 g/L
D(+)-glucose, 1 g/L
nalidixic acid, 0.04 g/L
sodium chloride, 5 g/L
thiamine dichloride, 0.005 g/L
Trypaflavin, 0.01 g/L
tryptose, 20 g/L

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

selective for Listeria spp.

Application

For the cultivation and isolation of Listeria from clinical and other material according to Feindt.

Other Notes

Isolation of L. monocytogenes with nalidixic acid and Trypaflavin[1][2]

Legal Information

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[Isolation of L. monocytogenes by nalidixic acid trypaflavine].
E H Kampelmacher et al.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Erste Abteilung Originale. Reihe A: Medizinische Mikrobiologie und Parasitologie, 221(1), 139-140 (1972-07-01)
G. Obiger et al.
Fleischwirtsch., 10, 1452-1452 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service