Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62351

Millipore

Listeria Selective Enrichment Supplement according to IDF/FIL

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

solid

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

Application

Supplement for Listeria Enrichment Broth acc. to IDF-FIL (Cat. No. 62353

Components

Composition: Nalidixic acid Sodium salt 20 mg, Cycloheximide 25 mg, Acriflavine hydrochloride 5 mg

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service