Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62348

Millipore

Listeria Selective Enrichment Supplement according to FDA

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

solid

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

Application

Supplement for Listeria Enrichment Broth acc. to. FDA (Cat. No. 62353)

Packaging

Set with 16 vials

Components

Acriflavine hydrochloride 7.5 mg, Cycloheximide 25 mg, Nalidixic acid Sodium salt 20 mg

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service