62112
3-Methylhexanal
analytical standard
Synonym(s):
3-Methylcapronaldehyde
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥97.0% (GC)
shelf life
limited shelf life, expiry date on the label
contains
~0.25% BHT as stabilizer
refractive index
n20/D 1.411
density
0.813 g/mL at 20 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
CCCC(C)CC([H])=O
InChI
1S/C7H14O/c1-3-4-7(2)5-6-8/h6-7H,3-5H2,1-2H3
InChI key
ZSJUABCTGCNBPF-UHFFFAOYSA-N
General description
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service