Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62108

Supelco

Lewatit® TP 260 disodium form

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41121813

description

weakly acidic cation exchange resin

Quality Level

form

beads

technique(s)

LPLC: suitable

loss

~60% loss on drying, 110 °C

matrix

styrene-divinylbenzene (macroporous)

matrix active group

(aminomethyl)phosphonic acid functional group

particle size

0.4-1.25 mm
20-50 mesh
400-1250 μm

capacity

2.3 meq/mL

separation technique

cation exchange

General description

Weakly acidic cation exchange resin with high available capacity.

Application

Chelating resin especially suited for refined purification of alkali chloride brines and for extraction/recycling of heavy metal ions.

Legal Information

Lewatit is a registered trademark of Lanxess Deutschland GmbH

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service