62093
Lewatit® MP-64 chloride form
anion exchange resin crosslinked polystyrene matrix, free base, medium-basic, macroporous
About This Item
Recommended Products
form
macroporous
moisture
50-65%
technique(s)
LPLC: suitable
loss
50-65% loss on drying, 110 °C
matrix
styrene divinylbenzene (macroporous)
matrix active group
tertiary and quaternary ammonium groups
particle size
300-1,250 μm
separation technique
anion exchange
General description
Application
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service