Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61923

Supelco

Bisphenol TMC

analytical standard

Synonym(s):

1,1-Bis(4-hydroxyphenyl)-3,3,5-trimethylcyclohexane, 4,4′-(3,3,5-Trimethylcyclohexylidene)bisphenol, 4,4′-(3,3,5-Trimethylcyclohexylidene)diphenol, BISP-TMC, BPTMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
Beilstein/REAXYS Number:
9785841
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC1CC(C)(C)CC(C1)(c2ccc(O)cc2)c3ccc(O)cc3

InChI

1S/C21H26O2/c1-15-12-20(2,3)14-21(13-15,16-4-8-18(22)9-5-16)17-6-10-19(23)11-7-17/h4-11,15,22-23H,12-14H2,1-3H3

InChI key

UMPGNGRIGSEMTC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service